ADV509448381 AChemBlock

3,3-Difluoropyrrolidin-2-one

  • MDL number MFCD13186732
  • Cas Number : 162970-49-2
  • Molecular Weight : 121.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-3-difluoropyrrolidin-2-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14522
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart