ADV509448382 AChemBlock

Methyl 5-bromo-4,6-difluoro-1H-indole-3-carboxylate

  • MDL number MFCD27987758
  • Cas Number : 1638763-46-8
  • Molecular Weight : 290.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromo-4-6-difluoro-1h-indole-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14523
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart