ADV509448384 AChemBlock

tert-Butyl N-[(2R,3R)-2-methylpiperidin-3-yl]carbamate

  • MDL number MFCD22398987
  • Cas Number : 1791402-58-8
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-2r-3r-2-methylpiperidin-3-yl-carbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14525
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart