ADV509448386 AChemBlock

(3S)-3-Ethylpyrrolidine hydrochloride

  • MDL number MFCD28501900
  • Cas Number : 235094-03-8
  • Molecular Weight : 135.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3s-3-ethylpyrrolidine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14527
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart