ADV509448390 AChemBlock

tert-Butyl N-[(2S,3S)-2-methylpiperidin-3-yl]carbamate

  • MDL number MFCD29918579
  • Cas Number : 1860012-45-8
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-2s-3s-2-methylpiperidin-3-yl-carbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14531
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart