ADV509448392 AChemBlock

6-Chloro-2-methyl-5-nitropyridine-3-carboxylic acid

  • MDL number MFCD27988107
  • Cas Number : 1638765-04-4
  • Molecular Weight : 216.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-2-methyl-5-nitropyridine-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14534
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart