ADV509448468 AChemBlock

2-Bromo-1-(3-bromo-4-methyl-phenyl)-ethanone

  • MDL number MFCD19345958
  • Cas Number : 60208-05-1
  • Molecular Weight : 291.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-1-3-bromo-4-methyl-phenyl-ethanone.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15182
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart