ADV509448481 AChemBlock

Ethyl 5-amino-1-phenyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD07388406
  • Cas Number : 866837-96-9
  • Molecular Weight : 231.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-5-amino-1-phenyl-1h-pyrazole-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15196
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart