ADV509448483 AChemBlock

2,4-Dichlorobenzofuro[3,2-d]pyrimidine

  • MDL number MFCD13190348
  • Cas Number : 160199-95-1
  • Molecular Weight : 239.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-4-dichlorobenzofuro-3-2-d-pyrimidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15198
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart