ADV509448509 AChemBlock

4-Bromo-7-fluoro-5-azaindole

  • MDL number MFCD11845516
  • Cas Number : 1082040-86-5
  • Molecular Weight : 215.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-7-fluoro-5-azaindole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15226
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart