ADV509448518 AChemBlock

5-Chloro-6-methyl-1H-indole

  • MDL number MFCD09026970
  • Cas Number : 162100-56-3
  • Molecular Weight : 165.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-6-methyl-1h-indole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15235
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart