ADV509448530 AChemBlock

4-Benzyloxy-2-oxindole

  • MDL number MFCD28101618
  • Cas Number : 458526-10-8
  • Molecular Weight : 239.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-benzyloxy-2-oxindole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15249
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart