ADV509448534 AChemBlock

4,6-Dichloro-5-azaindole

  • MDL number MFCD09038480
  • Cas Number : 67139-79-1
  • Molecular Weight : 187.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-6-dichloro-5-azaindole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15254
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart