ADV509448545 AChemBlock

6-Bromo-3-hydroxy-1H-indazole

  • MDL number MFCD07781600
  • Cas Number : 885521-92-6
  • Molecular Weight : 213.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-3-hydroxy-1h-indazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15267
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart