ADV509448551 AChemBlock

3-Methyl-2-(4-methylphenyl)morpholine

  • MDL number MFCD11646217
  • Cas Number : 1094649-71-4
  • Molecular Weight : 191.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-2-4-methylphenyl-morpholine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15274
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart