ADV509448570 AChemBlock

Gemigliptin (2R,3R)-2,3-dihydroxybutanedioate

  • MDL number MFCD19443742
  • Cas Number : 1374639-74-3
  • Molecular Weight : 639.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/gemigliptin-2r-3r-2-3-dihydroxybutanedioate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14550
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart