ADV509448574 AChemBlock

Ethyl 4-methoxy-5-methylpyrrolo[2,1-f][1,2,4]triazine-6-carboxylate

  • MDL number MFCD30146478
  • Cas Number : 1860028-32-5
  • Molecular Weight : 235.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-methoxy-5-methylpyrrolo-2-1-f-1-2-4-triazine-6-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14340
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart