ADV638390143 AChemBlock

5-(1-Hydroxy-1-methyl-ethyl)-pyridine-2-carboxylic acid

  • MDL number MFCD30502622
  • Cas Number : 2068151-91-5
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-1-hydroxy-1-methyl-ethyl-pyridine-2-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K12256
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart