ADV638390163 AChemBlock

Rifapentine

  • MDL number MFCD00866806
  • Cas Number : 61379-65-5
  • Molecular Weight : 877.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/rifapentine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15662
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart