ADV638390171 AChemBlock

1H,4H-pyrrolo[2,1-f][1,2,4]triazin-4-one

  • MDL number MFCD12923560
  • Cas Number : 159326-71-3
  • Molecular Weight : 135.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/pyrrolo-1-2-f-1-2-4-triazin-4-3h-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5644
pictograms GHS07
hstatements H302;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart