ADV638390237 AChemBlock

tert-Butyl 2-(fluoromethyl)-4-oxopiperidine-1-carboxylate

  • MDL number MFCD11975879
  • Cas Number : 1260674-52-9
  • Molecular Weight : 231.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-fluoromethyl-4-oxopiperidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8306
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart