ADV638390283 AChemBlock

6-Mercapto-nicotinic acid methyl ester

  • MDL number MFCD01860213
  • Cas Number : 74470-34-1
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-mercapto-nicotinic-acid-methyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8392
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart