ADV638390314 AChemBlock

cis-3-Hydroxy-3-methylcyclobutanecarboxylic acid

  • MDL number MFCD23106032
  • Cas Number : 1384855-41-7
  • Molecular Weight : 130.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/cis-3-hydroxy-3-methylcyclobutanecarboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9401
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart