ADV638390315 AChemBlock

tert-butyl 2-oxo-8-azaspiro[4.5]decane-8-carboxylate

  • MDL number MFCD18792443
  • Cas Number : 1250994-14-9
  • Molecular Weight : 253.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-boc-2-oxo-8-azaspiro-4-5-decane-8-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9569
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart