ADV638390530 AChemBlock

2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

  • MDL number MFCD13181964
  • Cas Number : 837392-54-8
  • Molecular Weight : 257.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1h-indole-2-methyl-5-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K10893
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart