ADV638390632 AChemBlock

2-(2-Nitrophenyl)acetaldehyde

  • MDL number MFCD08703363
  • Cas Number : 1969-73-9
  • Molecular Weight : 165.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Red liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-2-nitrophenyl-acetaldehyde.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8896
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart