ADV638390827 AChemBlock

[2-amino-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol

  • MDL number MFCD22494407
  • Cas Number : 1339927-45-5
  • Molecular Weight : 249.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-amino-5-tetramethyl-1-3-2-dioxaborolan-2-yl-phenyl-methanol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14575
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart