ADV638391016 AChemBlock

(1R,2R)-rel-2-(4-chloro-3-fluorophenyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD30502876
  • Cas Number : 2055841-07-9
  • Molecular Weight : 214.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/1r-2r-rel-2-4-chloro-3-fluorophenyl-cyclopropane-1-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=L14761
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart