ADV638391116 AChemBlock

2-amino-3-{1H-pyrrolo[2,3-b]pyridin-3-yl}propanoic acid

  • MDL number MFCD00037930
  • Cas Number : 7303-50-6
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-amino-3-1h-pyrrolo-2-3-b-pyridin-3-yl-propanoic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14861
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart