ADV638391665 AChemBlock

tert-butyl 5-bromo-1,2,3,6-tetrahydropyridine-1-carboxylate

  • MDL number MFCD28054768
  • Cas Number : 1415841-82-5
  • Molecular Weight : 262.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5-bromo-1-2-3-6-tetrahydropyridine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L18376
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart