ADV638391726 AChemBlock

6-Chloro-2,7-dimethyl-imidazo[1,2-b]pyridazine

  • MDL number MFCD27946128
  • Cas Number : 17412-20-3
  • Molecular Weight : 181.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-2-7-dimethyl-imidazo-1-2-b-pyridazine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K12073
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart