ADV638391846 AChemBlock

1-(4-methoxybenzyl)-1H-pyrazolo[3,4-b]pyridin-4-ol

  • MDL number MFCD20485436
  • Cas Number : 924909-16-0
  • Molecular Weight : 255.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-4-methoxybenzyl-1h-pyrazolo-3-4-b-pyridin-4-ol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15336
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart