ADV638391991 AChemBlock

4-Fluoro-indole-6-carbonitrile

  • MDL number MFCD23706316
  • Cas Number : 1427389-62-5
  • Molecular Weight : 160.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-indole-6-carbonitrile.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15492
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart