ADV638392102 AChemBlock

Methyl 4-bromoindole-2-carboxylate

  • MDL number MFCD04966956
  • Cas Number : 167479-13-2
  • Molecular Weight : 254.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-bromoindole-2-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15613
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart