ADV638392441 AChemBlock

4-(methylamino)pyrrolidin-2-one

  • MDL number MFCD28132784
  • Cas Number : 1195687-54-7
  • Molecular Weight : 114.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-methylamino-pyrrolidin-2-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L19088
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart