ADV638393739 AChemBlock

2-(1,3-benzothiazol-2-yl)-2,4,5,6-tetrahydrocyclopenta[c]pyrazol-3-amine

  • MDL number MFCD16631736
  • Molecular Weight : 256.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/2-1-3-benzothiazol-2-yl-2-4-5-6-tetrahydrocyclopenta-c-pyrazol-3-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=L20804
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart