ADV772121112 AChemBlock

2-Bromo-5-methyl-4,5,6,7-tetrahydro[1,3]thiazolo[5,4-c]pyridine

  • MDL number MFCD22035142
  • Cas Number : 143150-92-9
  • Molecular Weight : 233.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-5-methyl-4-5-6-7-tetrahydro-1-3-thiazolo-5-4-c-pyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7811
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart