ADV947321362 AChemBlock

2-Phenoxyacetaldehyde

  • MDL number MFCD00134414
  • Cas Number : 2120-70-9
  • Molecular Weight : 136.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-phenoxyacetaldehyde.html

Safety Information

signalword Warning
pcodes P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K12296
pictograms GHS07
hstatements H303;H320
Availability details for
Quantity
  • Availability
  • Add to Cart