ADV947321494 AChemBlock

2-{1H-Pyrrolo[2,3-b]pyridin-3-yl}acetic acid

  • MDL number MFCD10698203
  • Cas Number : 1912-42-1
  • Molecular Weight : 176.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-1h-pyrrolo-2-3-b-pyridin-3-yl-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L21587
Availability details for
Quantity
  • Availability
  • Add to Cart