ADV947321873 AChemBlock

Methyl 3-amino-1H-pyrazole-5-carboxylate

  • MDL number MFCD09991917
  • Cas Number : 632365-54-9
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-amino-1h-pyrazole-5-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-899
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart