ADV947321894 AChemBlock

5-Iodo-2-methyl-1,3-benzothiazole

  • MDL number MFCD00643249
  • Cas Number : 90414-61-2
  • Molecular Weight : 275.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Store at Room Tem.
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/5-iodo-2-methyl-1-3-benzothiazole.html

Safety Information

signalword Warning
pcodes P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L21650
pictograms GHS07
hstatements H303;H320
Availability details for
Quantity
  • Availability
  • Add to Cart