ADV947322810 AChemBlock

N-Phenylglycine tert-Butyl ester

  • MDL number MFCD11848799
  • Cas Number : 65171-67-7
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/n-phenylglycine-tert-butyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M23295
Availability details for
Quantity
  • Availability
  • Add to Cart