ADV947322822 AChemBlock

2-Ethynyl-5-fluoro-pyridine

  • MDL number MFCD03095259
  • Cas Number : 884494-34-2
  • Molecular Weight : 121.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-ethynyl-5-fluoro-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M22957
Availability details for
Quantity
  • Availability
  • Add to Cart