ADV947323045 AChemBlock

4'-Propyl-biphenyl-4-amine

  • MDL number MFCD12824134
  • Cas Number : 60040-12-2
  • Molecular Weight : 211.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-propyl-biphenyl-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M23183
Availability details for
Quantity
  • Availability
  • Add to Cart