ADV947323392 AChemBlock

4-Benzyloxypiperidine hydrochloride

  • MDL number MFCD07368306
  • Cas Number : 81151-68-0
  • Molecular Weight : 227.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-benzyloxypiperidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M22692
Availability details for
Quantity
  • Availability
  • Add to Cart