ADV947323395 AChemBlock

1-Boc-3-acetyl-piperidine

  • MDL number MFCD09953372
  • Cas Number : 858643-92-2
  • Molecular Weight : 227.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-acetyl-piperidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M22653
Availability details for
Quantity
  • Availability
  • Add to Cart