ADV947323405 AChemBlock

4-[4-(Trifluoromethoxy)phenyl]-piperidine hydrochloride

  • MDL number MFCD28109000
  • Cas Number : 180160-91-2
  • Molecular Weight : 281.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-4-trifluoromethoxy-phenyl-piperidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M22645
Availability details for
Quantity
  • Availability
  • Add to Cart