ADV947323433 AChemBlock

Methyl 3-amino-3-phenylpropanoate hydrochloride

  • MDL number MFCD00563398
  • Cas Number : 88831-43-0
  • Molecular Weight : 215.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-amino-3-phenylpropanoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M22874
Availability details for
Quantity
  • Availability
  • Add to Cart