ADV947323437 AChemBlock

Methyl 3-amino-3-(3-bromophenyl)propanoate hydrochloride

  • MDL number MFCD13195917
  • Cas Number : 1038381-80-4
  • Molecular Weight : 294.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-amino-3-3-bromophenyl-propanoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M23059
Availability details for
Quantity
  • Availability
  • Add to Cart