ADV998067126 AChemBlock

5-Bromo-4-chloro-pyridine-3-carbaldehyde

  • MDL number MFCD13189052
  • Cas Number : 1060802-24-5
  • Molecular Weight : 220.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-4-chloro-pyridine-3-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=N26075
Availability details for
Quantity
  • Availability
  • Add to Cart